accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 86217-82-5

H3CNH2

CAS RN 86217-82-5

IUPAC Name: 1-{[1,1'-biphenyl]-4-yl}ethan-1-amine
Synonyms: 1-{[1,1'-biphenyl]-4-yl}ethanamine
Molecular Weight (g/mol): 197.28
Molecular Formula: C14H15N
InChi Key: QVLZRPPCCDKMPO-UHFFFAOYNA-N
SMILES: CC(N)C1=CC=C(C=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results