accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NBOOCH3CH3CH3CH3

CAS RN 859833-21-9

IUPAC Name: 1-{[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}piperidine
Synonyms: 1-{[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}piperidine
Molecular Weight (g/mol): 301.24
Molecular Formula: C18H28BNO2
InChi Key: KEKNXVTYDJCRMT-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(CN2CCCCC2)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results