accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 859833-13-9

IUPAC Name: 2,4-dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-thiazole
Synonyms: 2,4-dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-thiazole
Molecular Weight (g/mol): 239.14
Molecular Formula: C11H18BNO2S
InChi Key: AZYDPQHPHNHZPL-UHFFFAOYSA-N
SMILES: CC1=NC(C)=C(S1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results