accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 858676-60-5

NI

CAS RN 858676-60-5

IUPAC Name: 1-[(4-iodophenyl)methyl]pyrrolidine
Synonyms: 1-[(4-iodophenyl)methyl]pyrrolidine
Molecular Weight (g/mol): 287.14
Molecular Formula: C11H14IN
InChi Key: RDHZJYCRHTZBIX-UHFFFAOYSA-N
SMILES: IC1=CC=C(CN2CCCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results