accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 857284-27-6

H3CNHO

CAS RN 857284-27-6

IUPAC Name: {[3-(furan-2-yl)phenyl]methyl}(methyl)amine
Synonyms: {[3-(furan-2-yl)phenyl]methyl}(methyl)amine
Molecular Weight (g/mol): 187.24
Molecular Formula: C12H13NO
InChi Key: WMCXJYGZLCREMM-UHFFFAOYSA-N
SMILES: CNCC1=CC=CC(=C1)C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results