accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NSOH

CAS RN 857283-96-6

IUPAC Name: [4-(2-methyl-1,3-thiazol-4-yl)phenyl]methanol
Synonyms: [4-(2-methyl-1,3-thiazol-4-yl)phenyl]methanol
Molecular Weight (g/mol): 205.28
Molecular Formula: C11H11NOS
InChi Key: RJHSPNFHEAFWDW-UHFFFAOYSA-N
SMILES: CC1=NC(=CS1)C1=CC=C(CO)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results