accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NNH

CAS RN 857283-76-2

IUPAC Name: N-{[4-(piperidin-1-yl)phenyl]methyl}cyclopropanamine
Synonyms: N-{[4-(piperidin-1-yl)phenyl]methyl}cyclopropanamine
Molecular Weight (g/mol): 230.36
Molecular Formula: C15H22N2
InChi Key: UNFZQBXRXGLJLF-UHFFFAOYSA-N
SMILES: C(NC1CC1)C1=CC=C(C=C1)N1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results