accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 855763-77-8

H3COONCH3

CAS RN 855763-77-8

IUPAC Name: ethyl 2-methylquinoline-6-carboxylate
Synonyms: ethyl 2-methylquinoline-6-carboxylate
Molecular Weight (g/mol): 215.25
Molecular Formula: C13H13NO2
InChi Key: WLKKUWXLOSMSMH-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C2N=C(C)C=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results