accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OHN

CAS RN 85553-55-5

IUPAC Name: [3-(pyridin-4-yl)phenyl]methanol
Synonyms: [3-(pyridin-4-yl)phenyl]methanol
Molecular Weight (g/mol): 185.23
Molecular Formula: C12H11NO
InChi Key: ZORHPGKUTIFODF-UHFFFAOYSA-N
SMILES: OCC1=CC=CC(=C1)C1=CC=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results