CAS RN 853771-88-7
IUPAC Name:
4-bromo-1-methoxy-2-(trifluoromethoxy)benzene
Synonyms:
4-bromo-1-methoxy-2-(trifluoromethoxy)benzene
Molecular Weight (g/mol):
271.03
Molecular Formula:
C8H6BrF3O2
InChi Key:
QVGIROOBNUFIKC-UHFFFAOYSA-N
SMILES:
COC1=C(OC(F)(F)F)C=C(Br)C=C1
1
–
1
of
1
results
1
4-Bromo-2-(trifluoromethoxy)anisole, 98%
CAS: 853771-88-7 Molecular Formula: C8H6BrF3O2 Molecular Weight (g/mol): 271.03 MDL Number: MFCD09907950 InChI Key: QVGIROOBNUFIKC-UHFFFAOYSA-N Synonym: 4-bromo-2-trifluoromethoxy anisole,4-bromo-1-methoxy-2-trifluoromethoxy benzene,benzene,4-bromo-1-methoxy-2-trifluoromethoxy PubChem CID: 45790939 IUPAC Name: 4-bromo-1-methoxy-2-(trifluoromethoxy)benzene SMILES: COC1=C(OC(F)(F)F)C=C(Br)C=C1