accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NNO

CAS RN 852227-92-0

IUPAC Name: 3-(1H-pyrazol-1-yl)benzaldehyde
Synonyms: 3-(pyrazol-1-yl)benzaldehyde
Molecular Weight (g/mol): 172.19
Molecular Formula: C10H8N2O
InChi Key: NKFXXJOWQSOGOF-UHFFFAOYSA-N
SMILES: O=CC1=CC(=CC=C1)N1C=CC=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results