accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 852180-76-8

ONOOH

CAS RN 852180-76-8

IUPAC Name: {4-[2-(morpholin-4-yl)ethoxy]phenyl}methanol
Synonyms: {4-[2-(morpholin-4-yl)ethoxy]phenyl}methanol
Molecular Weight (g/mol): 237.3
Molecular Formula: C13H19NO3
InChi Key: LZRLXLXSGAEQLA-UHFFFAOYSA-N
SMILES: OCC1=CC=C(OCCN2CCOCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results