accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OOOOH

CAS RN 851048-51-6

IUPAC Name: 4-(oxan-4-yloxy)benzoic acid
Synonyms: 4-(oxan-4-yloxy)benzoic acid
Molecular Weight (g/mol): 222.24
Molecular Formula: C12H14O4
InChi Key: MPNYLYRGNCZFFH-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(OC2CCOCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results