accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NNH3CBr

CAS RN 85100-77-2

IUPAC Name: 1-butyl-3-methyl-1H-imidazol-3-ium bromide
Synonyms: 1-butyl-3-methylimidazolium bromide
Molecular Weight (g/mol): 219.13
Molecular Formula: C8H15BrN2
InChi Key: KYCQOKLOSUBEJK-UHFFFAOYSA-M
SMILES: [Br-].CCCCN1C=C[N+](C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results