accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 85070-47-9

FClBr

CAS RN 85070-47-9

IUPAC Name: 1-(bromomethyl)-3-chloro-2-fluorobenzene
Synonyms: 1-(bromomethyl)-3-chloro-2-fluorobenzene
Molecular Weight (g/mol): 223.47
Molecular Formula: C7H5BrClF
InChi Key: HYILLTADABKYHO-UHFFFAOYSA-N
SMILES: FC1=C(Cl)C=CC=C1CBr
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)