accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 850593-10-1

BFFFOCH3HOOH

CAS RN 850593-10-1

IUPAC Name: [2-ethoxy-5-(trifluoromethyl)phenyl]boronic acid
Synonyms: 2-ethoxy-5-(trifluoromethyl)phenylboronic acid
Molecular Weight (g/mol): 233.98
Molecular Formula: C9H10BF3O3
InChi Key: IOCXELVDHLDNMQ-UHFFFAOYSA-N
SMILES: CCOC1=C(C=C(C=C1)C(F)(F)F)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results