accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 850593-06-5

BFCH3HOOH

CAS RN 850593-06-5

IUPAC Name: (3-fluoro-5-methylphenyl)boronic acid
Synonyms: 3-fluoro-5-methylphenylboronic acid
Molecular Weight (g/mol): 153.95
Molecular Formula: C7H8BFO2
InChi Key: QQPLHUGOUZKARP-UHFFFAOYSA-N
SMILES: CC1=CC(F)=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results