accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

BOOH3CCH3H3CH3CNH2HCl

CAS RN 850568-55-7

IUPAC Name: 1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanamine hydrochloride
Synonyms: 1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanamine hydrochloride
Molecular Weight (g/mol): 269.58
Molecular Formula: C13H21BClNO2
InChi Key: KPECMJIHZZWTJN-UHFFFAOYSA-N
SMILES: Cl.CC1(C)OB(OC1(C)C)C1=CC=C(CN)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results