accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

BHNOCH3HOOH

CAS RN 850568-41-1

IUPAC Name: [4-(acetamidomethyl)phenyl]boronic acid
Synonyms: 4-(acetamidomethyl)phenylboronic acid
Molecular Weight (g/mol): 193.01
Molecular Formula: C9H12BNO3
InChi Key: ZMJVNKSOLIUBKO-UHFFFAOYSA-N
SMILES: CC(=O)NCC1=CC=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results