accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 850568-05-7

CH3OOFBOHHO

CAS RN 850568-05-7

IUPAC Name: [5-fluoro-2-(methoxycarbonyl)phenyl]boronic acid
Synonyms: 5-fluoro-2-(methoxycarbonyl)phenylboronic acid
Molecular Weight (g/mol): 197.96
Molecular Formula: C8H8BFO4
InChi Key: CHDCMVDJRAQCTJ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(F)C=C1B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results