accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 850374-94-6

H3CH3CCH3OONOOCH3I

CAS RN 850374-94-6

IUPAC Name: 1-tert-butyl 6-methyl 3-iodo-1H-indole-1,6-dicarboxylate
Synonyms: 1-tert-butyl 6-methyl 3-iodoindole-1,6-dicarboxylate
Molecular Weight (g/mol): 401.2
Molecular Formula: C15H16INO4
InChi Key: KFPCBJZNHXTGHX-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C2C(I)=CN(C(=O)OC(C)(C)C)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results