accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 85017-77-2

CoBrBr

CAS RN 85017-77-2

IUPAC Name: dibromocobalt hydrate
Synonyms: cobalt(II) bromide hydrate
Molecular Weight (g/mol): 236.76
Molecular Formula: Br2CoH2O
InChi Key: MOTZAWYGLXXRSO-UHFFFAOYSA-L
SMILES: O.Br[Co]Br
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)