accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 85006-23-1

IUPAC Name: hydrogen (3-aminophenyl)boronic acid chloride
Synonyms: hydrogen M-aminophenylboronic acid chloride
Molecular Weight (g/mol): 173.4
Molecular Formula: C6H9BClNO2
InChi Key: QBMHZZHJIBUPOX-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC1=CC=CC(=C1)B(O)O
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results