accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3HNOOH3C

CAS RN 85-91-6

IUPAC Name: methyl 2-(methylamino)benzoate
Synonyms: methyl 2-(methylamino)benzoate
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: GVOWHGSUZUUUDR-UHFFFAOYSA-N
SMILES: CNC1=CC=CC=C1C(=O)OC
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)