accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OHSOON

CAS RN 85-48-3

IUPAC Name: quinoline-8-sulfonic acid
Synonyms: quinoline-8-sulfonic acid
Molecular Weight (g/mol): 209.22
Molecular Formula: C9H7NO3S
InChi Key: SXVRECLPTCOMIA-UHFFFAOYSA-N
SMILES: OS(=O)(=O)C1=C2N=CC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results