accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CNHSHCl

CAS RN 849776-43-8

IUPAC Name: [(1-benzothiophen-2-yl)methyl](methyl)amine hydrochloride
Synonyms: (1-benzothiophen-2-ylmethyl)(methyl)amine hydrochloride
Molecular Weight (g/mol): 213.72
Molecular Formula: C10H12ClNS
InChi Key: PBEZSHRKTUDPLK-UHFFFAOYSA-N
SMILES: Cl.CNCC1=CC2=CC=CC=C2S1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results