CAS RN 846-70-8
CAS RN 846-70-8
IUPAC Name:
sodium 2,4-dinitro-7-sulfonatonaphthalen-1-olate
Synonyms:
acid yellow 1
Molecular Weight (g/mol):
335.2
Molecular Formula:
C10H4N2NaO8S
InChi Key:
CGXSFOUXEQBDCJ-UHFFFAOYSA-L
SMILES:
[Na+].[O-]C1=C(C=C(C2=CC=C(C=C12)S([O-])(=O)=O)[N+]([O-])=O)[N+]([O-])=O
1
–
1
of
1
results
1
Naphthol Yellow S, Thermo Scientific™
CAS: 846-70-8 Molecular Formula: C10H4N2Na2O8S Molecular Weight (g/mol): 358.188 MDL Number: MFCD00003958 InChI Key: CTIQLGJVGNGFEW-UHFFFAOYSA-L Synonym: C.I. 10316; 5,7-Dinitro-8-hydroxy-2-naphthalenesulfonic acid disodium salt PubChem CID: 2724063 ChEBI: CHEBI:87219 IUPAC Name: disodium;5,7-dinitro-8-oxidonaphthalene-2-sulfonate SMILES: C1=CC2=C(C=C1S(=O)(=O)[O-])C(=C(C=C2[N+](=O)[O-])[N+](=O)[O-])[O-].[Na+].[Na+]