accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 845885-94-1

CH3NNNBrOOH

CAS RN 845885-94-1

IUPAC Name: 1-[(4-bromophenyl)methyl]-5-methyl-1H-1,2,3-triazole-4-carboxylic acid
Synonyms: 1-[(4-bromophenyl)methyl]-5-methyl-1,2,3-triazole-4-carboxylic acid
Molecular Weight (g/mol): 296.12
Molecular Formula: C11H10BrN3O2
InChi Key: FJFFHBYCFJNIAV-UHFFFAOYSA-N
SMILES: CC1=C(N=NN1CC1=CC=C(Br)C=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results