accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OHOOF

CAS RN 845885-90-7

IUPAC Name: 4-fluoro-3-formylbenzoic acid
Synonyms: 4-fluoro-3-formylbenzoic acid
Molecular Weight (g/mol): 168.12
Molecular Formula: C8H5FO3
InChi Key: SKPWEADPCJMLID-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(C=O)=C(F)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results