accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COONCH3CH3

CAS RN 84562-48-1

IUPAC Name: 4-(dimethylamino)-2-methoxybenzaldehyde
Synonyms: 4-(dimethylamino)-2-methoxybenzaldehyde
Molecular Weight (g/mol): 179.22
Molecular Formula: C10H13NO2
InChi Key: HGDRXADJVGVGBC-UHFFFAOYSA-N
SMILES: COC1=CC(=CC=C1C=O)N(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results