CAS RN 845551-44-2
CAS RN 845551-44-2
IUPAC Name:
[4-(benzyloxy)-3-chlorophenyl]boronic acid
Synonyms:
4-(benzyloxy)-3-chlorophenylboronic acid
Molecular Weight (g/mol):
262.5
Molecular Formula:
C13H12BClO3
InChi Key:
AYSMMNDQVZAXQY-UHFFFAOYSA-N
SMILES:
OB(O)C1=CC(Cl)=C(OCC2=CC=CC=C2)C=C1
1
–
1
of
1
results
1
4-Benzyloxy-3-chlorobenzeneboronic acid, 98%, Thermo Scientific™
CAS: 845551-44-2 Molecular Formula: C13H12BClO3 Molecular Weight (g/mol): 262.496 MDL Number: MFCD04115642 InChI Key: AYSMMNDQVZAXQY-UHFFFAOYSA-N Synonym: 4-benzyloxy-3-chlorophenylboronic acid,4-benzyloxy-3-chlorophenyl boronic acid,3-chloro-4-benzyloxyphenyl boronic acid,4-benzyloxy-3-chlorobenzeneboronic acid,boronic acid, 3-chloro-4-phenylmethoxy phenyl,b-3-chloro-4-phenylmethoxy phenyl-boronic acid,akos brn-0594,acmc-209pvk,4-benzyloxy-3-chlorophenylboro PubChem CID: 3763657 IUPAC Name: (3-chloro-4-phenylmethoxyphenyl)boronic acid SMILES: B(C1=CC(=C(C=C1)OCC2=CC=CC=C2)Cl)(O)O