accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 84547-60-4

CH3NNOCl

CAS RN 84547-60-4

IUPAC Name: 1-methyl-1H-pyrazole-3-carbonyl chloride
Synonyms: 1-methylpyrazole-3-carbonyl chloride
Molecular Weight (g/mol): 144.56
Molecular Formula: C5H5ClN2O
InChi Key: JCZIPPIUHKRMOM-UHFFFAOYSA-N
SMILES: CN1C=CC(=N1)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results