accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OOBr

CAS RN 844891-02-7

IUPAC Name: 1-(1-benzofuran-5-yl)-2-bromoethan-1-one
Synonyms: 1-(1-benzofuran-5-yl)-2-bromoethanone
Molecular Weight (g/mol): 239.07
Molecular Formula: C10H7BrO2
InChi Key: KRXJQVYCIGDILC-UHFFFAOYSA-N
SMILES: BrCC(=O)C1=CC=C2OC=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results