accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 84468-52-0

CH3NHHNCH3

CAS RN 84468-52-0

IUPAC Name: 2,3-dimethylpiperazine
Synonyms: 2,3-dimethylpiperazine
Molecular Weight (g/mol): 114.19
Molecular Formula: C6H14N2
InChi Key: COWPTMLRSANSMQ-UHFFFAOYNA-N
SMILES: CC1NCCNC1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results