accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3CH3H3COOHN(S)CH3OHOHO

CAS RN 84311-19-3

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-hydroxy-2-methylpropanoic acid
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]-3-hydroxy-2-methylpropanoic acid
Molecular Weight (g/mol): 219.24
Molecular Formula: C9H17NO5
InChi Key: FWRXDSRYWWYTPD-VIFPVBQESA-N
SMILES: CC(C)(C)OC(=O)N[C@@](C)(CO)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results