accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 84228-44-4

CH3OONH2Cl

CAS RN 84228-44-4

IUPAC Name: methyl 4-amino-3-chlorobenzoate
Synonyms: methyl 4-amino-3-chlorobenzoate
Molecular Weight (g/mol): 185.61
Molecular Formula: C8H8ClNO2
InChi Key: WKVJBYKFGNVWLM-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(N)C(Cl)=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)