accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 84192-88-1

H3COHNOOCH3CH3H3COHO

CAS RN 84192-88-1

IUPAC Name: 5-(tert-butoxy)-2-acetamido-5-oxopentanoic acid
Synonyms: 5-(tert-butoxy)-2-acetamido-5-oxopentanoic acid
Molecular Weight (g/mol): 245.28
Molecular Formula: C11H19NO5
InChi Key: FALCCLKESWGSNI-UHFFFAOYNA-N
SMILES: CC(=O)NC(CCC(=O)OC(C)(C)C)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)