accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CCH3(S)OOHNCH3OO

CAS RN 84000-11-3

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}(methyl)amino)-3-methylbutanoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl](methyl)amino}-3-methylbutanoic acid
Molecular Weight (g/mol): 353.42
Molecular Formula: C21H23NO4
InChi Key: YCXXXPZNQXXRIG-UHFFFAOYNA-N
SMILES: CC(C)C(N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)