accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NH2H2NCH3

CAS RN 84-67-3

IUPAC Name: 2,2'-dimethyl-[1,1'-biphenyl]-4,4'-diamine
Synonyms: 2,2'-dimethyl-[1,1'-biphenyl]-4,4'-diamine
Molecular Weight (g/mol): 212.3
Molecular Formula: C14H16N2
InChi Key: QYIMZXITLDTULQ-UHFFFAOYSA-N
SMILES: CC1=CC(N)=CC=C1C1=CC=C(N)C=C1C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)