accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 833482-53-4

CH3NHOF

CAS RN 833482-53-4

IUPAC Name: {[2-(4-fluorophenoxy)phenyl]methyl}(methyl)amine
Synonyms: {[2-(4-fluorophenoxy)phenyl]methyl}(methyl)amine
Molecular Weight (g/mol): 231.27
Molecular Formula: C14H14FNO
InChi Key: BSDBBIKNDVSUNF-UHFFFAOYSA-N
SMILES: CNCC1=CC=CC=C1OC1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results