accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 832114-06-4

BOOH3CCH3H3CH3CONNH3C

CAS RN 832114-06-4

IUPAC Name: 1-methyl-4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl]piperazine
Synonyms: 1-methyl-4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl]piperazine
Molecular Weight (g/mol): 330.24
Molecular Formula: C18H27BN2O3
InChi Key: TXCDVBDHDQAWLR-UHFFFAOYSA-N
SMILES: CN1CCN(CC1)C(=O)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)