accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 82771-60-6

IUPAC Name: 7-chloro-1,2,3,4-tetrahydroisoquinoline
Synonyms: 7-chloro-1,2,3,4-tetrahydroisoquinoline
Molecular Weight (g/mol): 167.64
Molecular Formula: C9H10ClN
InChi Key: BLRKQLICTKRDDZ-UHFFFAOYSA-N
SMILES: ClC1=CC2=C(CCNC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results