accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 82749-70-0

H3COClClOOOH

CAS RN 82749-70-0

IUPAC Name: 4-[(2-butyl-6,7-dichloro-2-cyclopentyl-1-oxo-2,3-dihydro-1H-inden-5-yl)oxy]butanoic acid
Synonyms: 4-[(2-butyl-6,7-dichloro-2-cyclopentyl-1-oxo-3H-inden-5-yl)oxy]butanoic acid
Molecular Weight (g/mol): 427.36
Molecular Formula: C22H28Cl2O4
InChi Key: KHKGTPJPBOQECW-UHFFFAOYNA-N
SMILES: CCCCC1(CC2=CC(OCCCC(O)=O)=C(Cl)C(Cl)=C2C1=O)C1CCCC1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (2)
  • (4)
  • (4)