accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NaOONOO

CAS RN 827-95-2

IUPAC Name: sodium 3-nitrobenzoate
Synonyms: sodium 3-nitrobenzoate
Molecular Weight (g/mol): 189.1
Molecular Formula: C7H4NNaO4
InChi Key: MUADFEZFSKAZLT-UHFFFAOYSA-M
SMILES: [Na+].[O-]C(=O)C1=CC=CC(=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results