accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 826-34-6

IUPAC Name: 1,2-dimethyl (1R,2S)-cyclopropane-1,2-dicarboxylate
Synonyms: 1,2-dimethyl (1R,2S)-cyclopropane-1,2-dicarboxylate
Molecular Weight (g/mol): 158.15
Molecular Formula: C7H10O4
InChi Key: JBVOSZYUSFDYIN-SYDPRGILSA-N
SMILES: COC(=O)[C@H]1C[C@H]1C(=O)OC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results