accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 824982-41-4

NONHSOOFFFNFFF

CAS RN 824982-41-4

IUPAC Name: N-(4-trifluoromethanesulfonylphenyl)-3'-(trifluoromethyl)-3,6-dihydro-2H-[1,2'-bipyridine]-4-carboxamide
Synonyms: N-(4-trifluoromethanesulfonylphenyl)-3'-(trifluoromethyl)-3,6-dihydro-2H-[1,2'-bipyridine]-4-carboxamide
Molecular Weight (g/mol): 479.4
Molecular Formula: C19H15F6N3O3S
InChi Key: SDUAWRFBHRAFBM-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=C(N=CC=C1)N1CCC(=CC1)C(=O)NC1=CC=C(C=C1)S(=O)(=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results