accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 82228-89-5

CH3NOSOOO

CAS RN 82228-89-5

IUPAC Name: 4-formyl-1-methylpyridin-1-ium benzenesulfonate
Synonyms: 4-formyl-1-methylpyridin-1-ium benzenesulfonate
Molecular Weight (g/mol): 279.31
Molecular Formula: C13H13NO4S
InChi Key: HSVLGIFAXFDLMU-UHFFFAOYSA-M
SMILES: C[N+]1=CC=C(C=O)C=C1.[O-]S(=O)(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results