accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 821-41-0

HOCH2

CAS RN 821-41-0

IUPAC Name: hex-5-en-1-ol
Synonyms: hex-5-en-1-ol
Molecular Weight (g/mol): 100.16
Molecular Formula: C6H12O
InChi Key: UIZVMOZAXAMASY-UHFFFAOYSA-N
SMILES: OCCCCC=C
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)