accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 821-38-5

HOOOHO

CAS RN 821-38-5

IUPAC Name: tetradecanedioic acid
Synonyms: tetradecanedioic acid
Molecular Weight (g/mol): 258.36
Molecular Formula: C14H26O4
InChi Key: HQHCYKULIHKCEB-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCCCCCCCCC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results