accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NNHO

CAS RN 82090-52-6

IUPAC Name: {imidazo[1,2-a]pyridin-2-yl}methanol
Synonyms: imidazo[1,2-a]pyridin-2-ylmethanol
Molecular Weight (g/mol): 148.17
Molecular Formula: C8H8N2O
InChi Key: ROPHYEIJSUUKEO-UHFFFAOYSA-N
SMILES: OCC1=CN2C=CC=CC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results